• USA Home
  • Search Results

  • hepessolution12345736545911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

HEPES solution

2 Products
Synonym:   N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)
Image of HEPES solution
CAS Number:
 MDL number:
 NACRES:
 NA.75
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Physical Form
  pH Value
  Application(s)
  Sterilization
Show More