• USA Home
  • Search Results

  • heptadecafluoro1iodooctane5459650763111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Heptadecafluoro-1-iodooctane

1 Products
Synonym:   1-Iodoperfluorooctane, Heptadecafluorooctyl iodide, Perfluorooctyl iodide
Image of Heptadecafluoro-<WBR>1-<WBR>iodooctane
CAS Number:
Linear Formula:
 CF3(CF2)7I
Molecular Weight:
 545.96
 Beilstein/REAXYS Number:
 1717141
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More