• USA Home
  • Search Results

  • heptadecafluorooctanesulfonicacidpotassiumsalt53822279539311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Heptadecafluorooctanesulfonic acid potassium salt

2 Products
Synonym:   Perfluorooctanesulfonic acid potassium salt, Potassium heptadecafluoro-1-octanesulfonate
Image of Heptadecafluorooctanesulfonic acid potassium salt
CAS Number:
Linear Formula:
 CF3(CF2)7SO3K
Molecular Weight:
 538.22
 Beilstein/REAXYS Number:
 3864579
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Special Grade
  Application(s)
Show More