• USA Home
  • Search Results

  • heptaldehyde1141911171711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Heptaldehyde

2 Products
Synonym:   1-Heptanal, Aldehyde C7, Enanthaldehyde, Oenanthaldehyde
Image of Heptaldehyde
CAS Number:
Linear Formula:
 CH3(CH2)5CHO
Molecular Weight:
 114.19
 Beilstein Registry Number:
 1560236
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More