• USA Home
  • Search Results

  • heptanal1141911171711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Heptanal

1 Products
Synonym:   1-Heptanal, Aldehyde C7, Enanthaldehyde, Heptaldehyde, Oenanthaldehyde
Image of Heptanal
CAS Number:
Linear Formula:
 CH3(CH2)5CHO
Molecular Weight:
 114.19
 Beilstein/REAXYS Number:
 1560236
 Council of Europe no.:
 117
 EC Number:
 FEMA Number:
 2540
 Flavis number:
 5.031
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More