• USA Home
  • Search Results

  • heptane1002014282511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Heptane

15 Products
Image of Heptane
CAS Number:
Linear Formula:
 CH3(CH2)5CH3
Molecular Weight:
 100.20
 Beilstein/REAXYS Number:
 1730763
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  UV-Absorption Values
  Agency/Method
  Anal. Standard Form
  Shelf Life
  Package quantity
  Restriction
  Suitability
  Purification method
  Absorption
  Transition temperature
  Product Line
  Ice Shipping
  Color
  UV-Absorption Condition
  Purity Grade
  Refractive Index
  Application(s)
  Greener Alternative Principles
  Physical Form
  Solubility
  Special Grade
  Assay
Show More