• USA Home
  • Search Results

  • heptanesmixtureofisomers1002014282511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Heptanes, mixture of isomers

1 Products
Image of Heptanes, mixture of isomers
CAS Number:
Empirical Formula (Hill Notation):
 C7H16
Molecular Weight:
 100.20
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Product Line
  Physical Form
Show More