• USA Home
  • Search Results

  • heptanoicacid1301811114811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Heptanoic acid

6 Products
Synonym:   Enanthic acid, Oenanthic acid
Image of Heptanoic acid
CAS Number:
Linear Formula:
 CH3(CH2)5COOH
Molecular Weight:
 130.18
 Beilstein/REAXYS Number:
 1744723
 Council of Europe no.:
 28
 EC Number:
 FEMA Number:
 3348
 Flavis number:
 8.028
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Refractive Index
  Document Links
  Special Grade
  Allergen
  Organoleptic property
  Titertest
  Solubility
  Assay
Show More