• USA Home
  • Search Results

  • heptylalcohol1162011170611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Heptyl alcohol

2 Products
Synonym:   1-Heptanol, Heptyl alcohol
Image of Heptyl alcohol
CAS Number:
Linear Formula:
 CH3(CH2)6OH
Molecular Weight:
 116.20
 Beilstein/REAXYS Number:
 1731686
 Council of Europe no.:
 70
 EC Number:
 FEMA Number:
 2548
 Flavis number:
 2.021
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Assay
  Agency/Method
Show More