• USA Home
  • Search Results

  • heptylamine1152211168211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Heptylamine

1 Products
Synonym:   1-Aminoheptane
Image of Heptylamine
CAS Number:
Linear Formula:
 CH3(CH2)6NH2
Molecular Weight:
 115.22
 Beilstein/REAXYS Number:
 1731688
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
126802 Heptylamine, 99%,