• USA Home
  • Search Results

  • heroind9solution37847133871349711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Heroin-D9 solution

2 Products
Image of Heroin-D<SUB>9</SUB> solution
CAS Number:
Empirical Formula (Hill Notation):
 C21H14NO5D9
Molecular Weight:
 378.47
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Anal. Standard Form
  Special Grade
  Package quantity
  Manufacturer designation
  Feature
Show More