• USA Home
  • Search Results

  • hesperidin6105652026311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hesperidin

6 Products
Synonym:   3′,5,7-Trihydroxy 4′-methoxyflavanone 7-rutinoside, Hesperetin 7-rhamnoglucoside, Hesperitin-7-rutinoside
Image of Hesperidin
CAS Number:
Empirical Formula (Hill Notation):
 C28H34O15
Molecular Weight:
 610.56
 Beilstein/REAXYS Number:
 75140
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Anal. Standard Form
  Physical Form
  Manufacturer designation
  Assay
  Application(s)
Show More