• USA Home
  • Search Results

  • hexachlorocyclopentadiene272777747411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexachlorocyclopentadiene

1 Products
Image of Hexachlorocyclopentadiene
CAS Number:
Empirical Formula (Hill Notation):
 C5Cl6
Molecular Weight:
 272.77
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More