• USA Home
  • Search Results

  • hexachloroethane236746772111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexachloroethane

2 Products
Synonym:   Perchloroethane
Image of Hexachloroethane
CAS Number:
Linear Formula:
 Cl3CCCl3
Molecular Weight:
 236.74
 Beilstein/REAXYS Number:
 1740341
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39050109
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Anal. Standard Form
  Physical Form
  Special Grade
  Assay
  Application(s)
Show More