• USA Home
  • Search Results

  • hexadecafluoroheptane3880533557911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexadecafluoroheptane

1 Products
Synonym:   Perfluoroheptane
Image of Hexadecafluoroheptane
CAS Number:
Linear Formula:
 CF3(CF2)5CF3
Molecular Weight:
 388.05
 Beilstein/REAXYS Number:
 1716335
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More