• USA Home
  • Search Results

  • hexadecane2264454476311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexadecane

3 Products
Synonym:   n-Hexadecane, Cetane
Image of Hexadecane
CAS Number:
Linear Formula:
 CH3(CH2)14CH3
Molecular Weight:
 226.44
 Beilstein/REAXYS Number:
 1736592
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Anal. Standard Form
  Application(s)
  Physical Form
  Special Grade
Show More