• USA Home
  • Search Results

  • hexadecylamine2414614327111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexadecylamine

3 Products
Synonym:   1-Aminohexadecane, Cetylamine, HAD
Image of Hexadecylamine
CAS Number:
Linear Formula:
 CH3(CH2)15NH2
Molecular Weight:
 241.46
 Beilstein/REAXYS Number:
 1634065
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More