• USA Home
  • Search Results

  • hexadecyltrimethylammoniumbromide364455709011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexadecyltrimethylammonium bromide

10 Products
Synonym:   CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide
Image of Hexadecyltrimethylammonium bromide
CAS Number:
Linear Formula:
 CH3(CH2)15N(Br)(CH3)3
Molecular Weight:
 364.45
 Beilstein/REAXYS Number:
 3598189
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Suitability
  UV-Absorption Condition
  Product Line
  Miscellaneous Specifications
  HLB Number
  Special Grade
  UV-Absorption Values
  pH Value
  ID clarifier
  Critical micelle concentration
  Application(s)
  Molecular weight
  Assay
  Physical Form
Show More

Product #
Description
H5882 Hexadecyltrimethylammonium bromide, ≥98%,
H6269 Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%,
H9151 Hexadecyltrimethylammonium bromide, BioXtra, ≥99%,
52365 Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT),
ARK2188 Hexadecyltrimethylammonium bromide, PharmaGrade, USP/NF, Manufactured under appropriate controls for pharma or biopharmaceutical production.,
52370 Hexadecyltrimethylammonium bromide, ≥96.0% (AT),
52367 Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur,
8.14119 Hexadecyltrimethylammonium bromide, for synthesis,
36932 Hexadecyltrimethylammonium bromide, analytical standard,
07883 Hexadecyltrimethylammonium bromide, puriss. p.a., matrix substance for MALDI-MS,