• USA Home
  • Search Results

  • hexaethyleneglycolmonotetradecylether47870515704011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexaethylene glycol monotetradecyl ether

1 Products
Synonym:   C14E6, Tetradecylhexaglycol
Image of Hexaethylene glycol monotetradecyl ether
CAS Number:
Linear Formula:
 CH3(CH2)13(OCH2CH2)6OH
Molecular Weight:
 478.70
 Beilstein/REAXYS Number:
 1805220
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More