• USA Home
  • Search Results

  • hexafluoroacetoneazine32806161984711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexafluoroacetone azine

1 Products
Image of Hexafluoroacetone azine
CAS Number:
Linear Formula:
 (CF3)2C=NN=C(CF3)2
Molecular Weight:
 328.06
 Beilstein/REAXYS Number:
 1916266
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More