• USA Home
  • Search Results

  • hexafluoroacetylacetone20806152222111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexafluoroacetylacetone

1 Products
Synonym:   1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, 1,1,1,5,5,5-Hexafluoroacetylacetone
Image of Hexafluoroacetylacetone
CAS Number:
Linear Formula:
 CF3COCH2COCF3
Molecular Weight:
 208.06
 Beilstein/REAXYS Number:
 1790138
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More