• USA Home
  • Search Results

  • hexamethoniumchloride273296025311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexamethonium chloride

1 Products
Synonym:   Hexamethonium dichloride, N,N,N,N′,N′,N′-Hexamethyl-1,6-hexanediaminium dichloride, N,N,N,N′,N′,N′-Hexamethylhexamethylenediammonium dichloride
CAS Number:
Linear Formula:
 (CH3)3N(Cl)(CH2)6N(Cl)(CH3)3
Molecular Weight:
 273.29
 Beilstein/REAXYS Number:
 3714806
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More