• USA Home
  • Search Results

  • hexamethyldisiloxane1623810746011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexamethyldisiloxane

6 Products
Synonym:   2,2,4,4-Tetramethyl-3-oxa-2,4-disilapentane, HMDSO, 2,2,4,4-Tetramethyl-3-oxa-2,4-disilapentane, HMDSO
Image of Hexamethyldisiloxane
CAS Number:
Linear Formula:
 ((CH3)3Si)2O
Molecular Weight:
 162.38
 EC Index Number
 203-492-7
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More