• USA Home
  • Search Results

  • hexamethylenetetramine1401910097011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexamethylenetetramine

7 Products
Synonym:   Hexamethylenetetramine, Hexamine, Formin, Urotropin, Hexamethylenetetramine, Hexamine, Formin, Urotropin
Image of Hexamethylenetetramine
CAS Number:
Empirical Formula (Hill Notation):
 C6H12N4
Molecular Weight:
 140.19
 EC Index Number
 202-905-8
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Mode of Action
  Physical Form
  Titertest
  Loss
  Solubility
  Assay
Show More