• USA Home
  • Search Results

  • hexamethylphosphoramide1792068031911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexamethylphosphoramide

3 Products
Synonym:   HMPA, Hexamethylphosphoric acid triamide, Tris(dimethylamino)phosphine oxide
Image of Hexamethylphosphoramide
CAS Number:
Linear Formula:
 [(CH3)2N]3PO
Molecular Weight:
 179.20
 Beilstein/REAXYS Number:
 1099903
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More