• USA Home
  • Search Results

  • hexanal100166625111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexanal

4 Products
Synonym:   Aldehyde C6, Caproaldehyde, Hexyl aldehyde
Image of Hexanal
CAS Number:
Linear Formula:
 CH3(CH2)4CHO
Molecular Weight:
 100.16
 Beilstein/REAXYS Number:
 506198
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39021112
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Document Links
  Allergen
  Organoleptic property
  Special Grade
  Agency/Method
  Assay
Show More