• USA Home
  • Search Results

  • hexane861811054311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexane

13 Products
Synonym:   n-Hexane
Image of Hexane
CAS Number:
Linear Formula:
 CH3(CH2)4CH3
Molecular Weight:
 86.18
 Beilstein/REAXYS Number:
 1730733
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Suitability
  Package quantity
  Agency/Method
  Solubility
  Refractive Index
  UV-Absorption Condition
  Transmittance
  Application(s)
  Purity Grade
  Restriction
  Assay
  UV-Absorption Values
Show More