• USA Home
  • Search Results

  • hexanemixtureofisomers861810783511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexane, mixture of isomers

8 Products
Synonym:   Hexanes, Isohexane
Image of Hexane, mixture of isomers
CAS Number:
Empirical Formula (Hill Notation):
 C6H14
Molecular Weight:
 86.18
 Beilstein/REAXYS Number:
 1730733
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  UV-Absorption Values
  pH Value
  Transmittance
  Product Line
  Solubility
  Density
  UV-Absorption Condition
  ID clarifier
  Physical Form
  Color
  Assay
  Special Grade
Show More