• USA Home
  • Search Results

  • hexanoicacid1161614262111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexanoic acid

6 Products
Synonym:   Acid C6, Caproic acid
Image of Hexanoic acid
CAS Number:
Linear Formula:
 CH3(CH2)4COOH
Molecular Weight:
 116.16
 Beilstein/REAXYS Number:
 773837
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Refractive Index
  Document Links
  Allergen
  Application(s)
  Potency
  Organoleptic property
  Solubility
  Assay
  Agency/Method
Show More