• USA Home
  • Search Results

  • hexylacetate1442114292711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexyl acetate

5 Products
Synonym:   Acetic acid hexyl ester, Acetic acid hexyl ester
Image of Hexyl acetate
CAS Number:
Empirical Formula (Hill Notation):
 C8H16O2
Molecular Weight:
 144.21
 EC Index Number
 607-462-00-8
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Assay
  Special Grade
Show More