• USA Home
  • Search Results

  • hexylbutyrate17226263963611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexyl butyrate

4 Products
Synonym:   Butyric acid hexyl ester, Hexyl butanoate, Butyric acid hexyl ester, Hexyl butanoate
Image of Hexyl butyrate
CAS Number:
Empirical Formula (Hill Notation):
 C10H20O2
Molecular Weight:
 172.26
 EC Index Number
 220-136-6
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Assay
  Special Grade
Show More