• USA Home
  • Search Results

  • hexyleneglycol1181710741511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexylene glycol

6 Products
Synonym:   (±)-2-Methyl-2,4-pentanediol, MPD
Image of Hexylene glycol
CAS Number:
Linear Formula:
 CH3CH(OH)CH2C(CH3)2OH
Molecular Weight:
 118.17
 Beilstein/REAXYS Number:
 1098298
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Special Grade
  UV-Absorption Values
  Application(s)
  Refractive Index
  Product Line
  Assay
Show More