• USA Home
  • Search Results

  • hexylhexanoate20032637865011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexyl hexanoate

3 Products
Synonym:   Hexyl caproate
Image of Hexyl hexanoate
CAS Number:
Linear Formula:
 CH3(CH2)4CO2(CH2)5CH3
Molecular Weight:
 200.32
 Council of Europe no.:
 316
 EC Number:
 FEMA Number:
 2572
 Flavis number:
 9.066
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Assay
  Special Grade
Show More