• USA Home
  • Search Results

  • hexylisobutyrate17226234907711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexyl isobutyrate

2 Products
Synonym:   Hexyl-2-methylpropionate
Image of Hexyl isobutyrate
CAS Number:
Linear Formula:
 (CH3)2CHCO2(CH2)5CH3
Molecular Weight:
 172.26
 Council of Europe no.:
 646
 EC Number:
 FEMA Number:
 3172
 Flavis number:
 9.478
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Special Grade
  Allergen
  Organoleptic property
  Assay
Show More