• USA Home
  • Search Results

  • hexylpropionate15824244576311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hexyl propionate

2 Products
Image of Hexyl propionate
CAS Number:
Linear Formula:
 C2H5CO2(CH2)5CH3
Molecular Weight:
 158.24
 Council of Europe no.:
 420
 EC Number:
 FEMA Number:
 2576
 Flavis number:
 9.139
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Assay
  Special Grade
Show More