• USA Home
  • Search Results

  • hmdi26235512430111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

HMDI

1 Products
Synonym:   4,4′-Diisocyanato-methylenedicyclohexane, 4,4′-HMDI, 4,4′-Methylene-bis(cyclohexyl isocyanate), Bis(4-isocyanatocyclohexyl)methane
Image of HMDI
CAS Number:
Linear Formula:
 CH2(C6H10NCO)2
Molecular Weight:
 262.35
 Beilstein/REAXYS Number:
 2217800
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More

Product #
Description
33490 HMDI, analytical standard,