• USA Home
  • Search Results

  • holmium16493744060011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Holmium

31 Products
Synonym:   Holmium-165
Image of Holmium
CAS Number:
Empirical Formula (Hill Notation):
 Ho
Molecular Weight:
 164.93
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Weight
  Particle Dimension
  Composite Dimension
  Manufacturer designation
  Assay
Show More

Product #
Description
457957 Holmium, chips, 99.9% trace metals basis,
GF83227686 Holmium, foil, thickness 0.25 mm, size 10 × 26 mm, purity 99%,
GF95140414 Holmium, lump, 25 mm max. lump size, weight 100 g, purity 99%,
GF13422934 Holmium, foil, thickness 0.50 mm, size 25 × 25 mm, tolerance 0.2,
GF29233335 Holmium, foil, thickness 0.50 mm, size 50 × 50 mm, tolerance 0.2,
GF35600510 Holmium, rod, 2.0 mm diameter, length 50 mm, purity 99%,
GF42361129 Holmium, lump, 25 mm max. lump size, weight 50 g, purity 99%,
GF19613684 Holmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%,
GF85502640 Holmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%,
GF38519376 Holmium, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99%,
GF49265253 Holmium, foil, not light tested, 25x25mm, thickness 0.0125mm, as rolled, 99%,
GF78609640 Holmium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%,
GF34764666 Holmium, foil, not light tested, 50x50mm, thickness 0.0125mm, as rolled, 99%,
GF60938635 Holmium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%,
GF42613006 Holmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99%,
GF26625067 Holmium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99%,
GF29991673 Holmium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%,
GF78567654 Holmium, powder, 10g, max. particle size 500 micron, 99.9%,
GF31028543 Holmium, powder, 5g, max. particle size 500 micron, 99.9%,
GF68907011 Holmium, rod, 100mm, diameter 3.8mm, 99%,
GF28769541 Holmium, rod, 100mm, diameter 6.35mm, cast, 99%,
GF13376785 Holmium, rod, 200mm, diameter 3.8mm, 99%,
GF92551259 Holmium, rod, 50mm, diameter 3.8mm, 99%,
GF12230154 Holmium, rod, 50mm, diameter 6.35mm, cast, 99%,
GF39809863 Holmium, wire reel, 100mm, diameter 0.5mm, as drawn, 99.9%,
GF22997188 Holmium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%,
GF59427513 Holmium, wire reel, 200mm, diameter 0.5mm, as drawn, 99.9%,
GF74648223 Holmium, wire reel, 200mm, diameter 1.0mm, hard, 99.9%,
GF88732693 Holmium, wire reel, 50mm, diameter 0.5mm, as drawn, 99.9%,
GF07383977 Holmium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%,
693693 Holmium, foil, thickness 1 mm, 99.9% trace rare earth metals basis,