• USA Home
  • Search Results

  • homogentisicacid1681545113811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Homogentisic acid

3 Products
Synonym:   2,5-Dihydroxyphenylacetic acid
Image of Homogentisic acid
CAS Number:
Linear Formula:
 (HO)2C6H3CH2CO2H
Molecular Weight:
 168.15
 Beilstein/REAXYS Number:
 2692860
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Special Grade
Show More