• USA Home
  • Search Results

  • homoharringtonine545622683387411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Homoharringtonine

2 Products
Synonym:   Ceflatonin, Cephalotaxine 4-methyl (2R)-2-hydroxy-2-(4-hydroxy-4-methylpentyl)butanedioate, Myelostat, NSC 141633, Omacetaxine mepesuccinate
Image of Homoharringtonine
CAS Number:
Empirical Formula (Hill Notation):
 C29H39NO9
Molecular Weight:
 545.62
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More