• USA Home
  • Search Results

  • homovanillicacid1821730608111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Homovanillic acid

2 Products
Synonym:   4-Hydroxy-3-methoxyphenylacetic acid, HVA
Image of Homovanillic acid
CAS Number:
Linear Formula:
 HOC6H3(OCH3)CH2CO2H
Molecular Weight:
 182.17
 Beilstein/REAXYS Number:
 2213447
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Assay
Show More