• USA Home
  • Search Results

  • humicacid12345141593611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Humic acid

1 Products
Synonym:   PGS 10
CAS Number:
 EC Number:
 MDL number:
 NACRES
 NA.22
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
53680 Humic acid, technical,