• USA Home
  • Search Results

  • hydriodicacid127911003485211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydriodic acid

7 Products
Synonym:   Hydriotic acid
Image of Hydriodic acid
CAS Number:
Empirical Formula (Hill Notation):
 HI
Molecular Weight:
 127.91
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Quality clarifier
  Purity Grade
  Concentration
  Special Grade
  Additive
  Density
Show More