• USA Home
  • Search Results

  • hydrochloricacid3646764701011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydrochloric acid

7 Products
Image of Hydrochloric acid
CAS Number:
Empirical Formula (Hill Notation):
 HCl
Molecular Weight:
 36.46
 Beilstein/REAXYS Number:
 1098214
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Product Line
  Suitability
  Restriction
  Solubility
  Purity Grade
  Color
  Concentration
  Special Grade
Show More