• USA Home
  • Search Results

  • hydrochlorothiazide297745893511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydrochlorothiazide

6 Products
Synonym:   6-Chloro-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, 6-Chloro-7-sulfamyl-3,4-dihydro-1,2,4-benzothiadiazine 1,1-dioxide
Image of Hydrochlorothiazide
CAS Number:
Empirical Formula (Hill Notation):
 C7H8ClN3O4S2
Molecular Weight:
 297.74
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Titertest
  Special Grade
  Anal. Standard Form
Show More