• USA Home
  • Search Results

  • hydrocortisone17butyrate432551360967111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydrocortisone 17-butyrate

1 Products
Synonym:   11β,17,21-Trihydroxypregn-4-ene-3,20-dione 17-butyrate, Cortisol 17-butyrate
Image of Hydrocortisone 17-butyrate
CAS Number:
Empirical Formula (Hill Notation):
 C25H36O6
Molecular Weight:
 432.55
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
H5270 Hydrocortisone 17-butyrate,