• USA Home
  • Search Results

  • hydrocortisone21acetate404505003311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydrocortisone 21-acetate

2 Products
Synonym:   11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-acetate, 17-α-Hydroxycorticosterone acetate, 17-Hydroxycorticosterone 21-acetate, 21-Acetoxy-4-pregnene-11β,17α-diol-3,20-dione, 4-Pregnene-11β,17α,21-triol-3,20-dione 21-acetate
Image of Hydrocortisone 21-acetate
CAS Number:
Empirical Formula (Hill Notation):
 C23H32O6
Molecular Weight:
 404.50
 Beilstein/REAXYS Number:
 2066841
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Special Grade
Show More