• USA Home
  • Search Results

  • hydrofluoricacid2001766439311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydrofluoric acid

4 Products
Synonym:   HF
Image of Hydrofluoric acid
CAS Number:
Empirical Formula (Hill Notation):
 HF
Molecular Weight:
 20.01
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Product Line
  Restriction
  Purity Grade
  Agency/Method
  Concentration
  Physical Form
  Assay
Show More