• USA Home
  • Search Results

  • hydrogen202133374011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydrogen

1 Products
Image of Hydrogen
CAS Number:
Empirical Formula (Hill Notation):
 H2
Molecular Weight:
 2.02
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  ID clarifier
Show More

Product #
Description
295396 Hydrogen, ≥99.99%,