• USA Home
  • Search Results

  • hydrogenchlorideethanolsolution3646764701011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydrogen chloride – ethanol solution

1 Products
Synonym:   Hydrochloric acid in ethanol
Image of Hydrogen chloride – ethanol solution
CAS Number:
Linear Formula:
 HCl · CH3CH2OH
Molecular Weight:
 36.46
 Beilstein/REAXYS Number:
 1098214
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss
 38050202
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Restriction
Show More