• USA Home
  • Search Results

  • hydrogenchloridesolution3646764701011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydrogen chloride solution

7 Products
Image of Hydrogen chloride solution
CAS Number:
Empirical Formula (Hill Notation):
 HCl
Molecular Weight:
 36.46
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Reaction Suitability
  Special Grade
  Quality clarifier
  Density
Show More